Anti-mAChR M2 Rabbit Polyclonal Antibody [clone: P224]
Lieferant: Bioworld Technology
BS2781
BWRLBS2781EA 262 EUR
BWRLBS2781
Anti-mAChR M2 Rabbit Polyclonal Antibody [clone: P224]
Antikörper
Synthetic peptide, corresponding to amino acids 191-240 of Human mAChR M2.
mAChR M2 (P224) pAb detects endogenous levels of mAChR M2 protein.
Type: Primary
Antigen: mAChR M2
Clonality: Polyclonal
Clone: P224
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat