Anti-RUNX-2 Rabbit Polyclonal Antibody [clone: N278]

Lieferant: Bioworld Technology
BS2831
BWRLBS2831EA 262 EUR
BWRLBS2831
Anti-RUNX-2 Rabbit Polyclonal Antibody [clone: N278]
Antikörper
Synthetic peptide, corresponding to amino acids 246-297 of Human RUNX-2.

RUNX2 (N278) pAb detects endogenous levels of RUNX2 protein.

Type: Primary
Antigen: RUNX-2
Clonality: Polyclonal
Clone: N278
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR