Anti-Myotubularin Rabbit Polyclonal Antibody [clone: Y290]

Lieferant: Bioworld Technology
BS2971
BWRLBS2971EA 262 EUR
BWRLBS2971
Anti-Myotubularin Rabbit Polyclonal Antibody [clone: Y290]
Antikörper
Synthetic peptide, corresponding to amino acids 256-3111 of Human Myotubularin.

Myotubularin (Y290) pAb detects endogenous levels of Myotubularin protein.

Type: Primary
Antigen: Myotubularin
Clonality: Polyclonal
Clone: Y290
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR