Anti-hnRNP K Rabbit Polyclonal Antibody [clone: Y280]
Lieferant: Bioworld Technology
BS2997
BWRLBS2997EA 262 EUR
BWRLBS2997
Anti-hnRNP K Rabbit Polyclonal Antibody [clone: Y280]
Antikörper
Synthetic peptide, corresponding to amino acids 250-300 of Human hnRNP K.
hnRNP K (Y280) pAb detects endogenous levels of hnRNP K protein.
Type: Primary
Antigen: hnRNP K
Clonality: Polyclonal
Clone: Y280
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat