Anti-TPX2 Rabbit Polyclonal Antibody [clone: Q330]

Lieferant: Bioworld Technology
BS3016
BWRLBS3016EA 262 EUR
BWRLBS3016
Anti-TPX2 Rabbit Polyclonal Antibody [clone: Q330]
Antikörper
Synthetic peptide, corresponding to amino acids 300-345 of Human TPX2.

TPX2 (Q330) pAb detects endogenous levels of TPX2 protein.

Type: Primary
Antigen: TPX2
Clonality: Polyclonal
Clone: Q330
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR