Anti-CYP7B1 Rabbit Polyclonal Antibody [clone: Q127]

Lieferant: Bioworld Technology
BS3024
BWRLBS3024EA 262 EUR
BWRLBS3024
Anti-CYP7B1 Rabbit Polyclonal Antibody [clone: Q127]
Antikörper
Synthetic peptide, corresponding to amino acids 101-150 of Human CYP7B1.

CYP7B1 (Q127) pAb detects endogenous levels of CYP7B1 protein.

Type: Primary
Antigen: CYP7B1
Clonality: Polyclonal
Clone: Q127
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR