Anti-RPL36 Rabbit Polyclonal Antibody [clone: V93]

Lieferant: Bioworld Technology
BS3051
BWRLBS3051EA 262 EUR
BWRLBS3051
Anti-RPL36 Rabbit Polyclonal Antibody [clone: V93]
Antikörper
Synthetic peptide, corresponding to amino acids 60-110 of Human RPL36.

RPL36 (V93) pAb detects endogenous levels of RPL36 protein.

Type: Primary
Antigen: RPL36
Clonality: Polyclonal
Clone: V93
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR