Anti-ACOT8 Rabbit Polyclonal Antibody [clone: P172]

Lieferant: Bioworld Technology
BS3062
BWRLBS3062EA 262 EUR
BWRLBS3062
Anti-ACOT8 Rabbit Polyclonal Antibody [clone: P172]
Antikörper
Synthetic peptide, corresponding to amino acids 151-200 of Human ACOT8.

ACOT8 (P172) pAb detects endogenous levels of ACOT8 protein.

Type: Primary
Antigen: ACOT8
Clonality: Polyclonal
Clone: P172
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR