Anti-AKR1B1 Rabbit Polyclonal Antibody [clone: K275]

Lieferant: Bioworld Technology
BS3067
BWRLBS3067EA 262 EUR
BWRLBS3067
Anti-AKR1B1 Rabbit Polyclonal Antibody [clone: K275]
Antikörper
Synthetic peptide, corresponding to amino acids 240-290 of Human AKR1B1.

AKR1B1 (K275) pAb detects endogenous levels of AKR1B1 protein.

Type: Primary
Antigen: AKR1B1
Clonality: Polyclonal
Clone: K275
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR