Anti-DNAL1 Rabbit Polyclonal Antibody [clone: N161]
Lieferant: Bioworld Technology
BS3115
BWRLBS3115EA 262 EUR
BWRLBS3115
Anti-DNAL1 Rabbit Polyclonal Antibody [clone: N161]
Antikörper
Synthetic peptide, corresponding to amino acids 128-184 of Human DNAL1.
DNAL1 (N161) pAb detects endogenous levels of DNAL1 protein.
Type: Primary
Antigen: DNAL1
Clonality: Polyclonal
Clone: N161
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat