Anti-GPR18 Rabbit Polyclonal Antibody [clone: C172]

Lieferant: Bioworld Technology
BS3164
BWRLBS3164EA 262 EUR
BWRLBS3164
Anti-GPR18 Rabbit Polyclonal Antibody [clone: C172]
Antikörper
Synthetic peptide, corresponding to amino acids 140-190 of Human GPR18.

GPR18 (C172) pAb detects endogenous levels of GPR18 protein.

Type: Primary
Antigen: GPR18
Clonality: Polyclonal
Clone: C172
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR