Anti-mAChR M4 Rabbit Polyclonal Antibody [clone: P277]

Lieferant: Bioworld Technology
BS3236
BWRLBS3236EA 262 EUR
BWRLBS3236
Anti-mAChR M4 Rabbit Polyclonal Antibody [clone: P277]
Antikörper
Synthetic peptide, corresponding to amino acids 241-290 of Human mAChR M4.

mAChR M4 (P277) pAb detects endogenous levels of mAChR M4 protein

Type: Primary
Antigen: mAChR M4
Clonality: Polyclonal
Clone: P277
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR