Anti-TG1019 Rabbit Polyclonal Antibody [clone: R251]

Lieferant: Bioworld Technology
BS3237
BWRLBS3237EA 262 EUR
BWRLBS3237
Anti-TG1019 Rabbit Polyclonal Antibody [clone: R251]
Antikörper
Synthetic peptide, corresponding to amino acids 220-270 of Human TG1019.

TG1019 (R251) pAb detects endogenous levels of TG1019 protein.

Type: Primary
Antigen: TG1019
Clonality: Polyclonal
Clone: R251
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR