Anti-ErbB2/HER2 Rabbit Polyclonal Antibody [clone: I682]

Lieferant: Bioworld Technology
BS3252
BWRLBS3252EA 262 EUR
BWRLBS3252
Anti-ErbB2/HER2 Rabbit Polyclonal Antibody [clone: I682]
Antikörper
Synthetic peptide, corresponding to amino acids 651-700 of Human ErbB2/HER2.

ErbB2/HER2 (I682) pAb detects endogenous levels of ErbB2/HER2 protein.

Type: Primary
Antigen: ErbB2/HER2
Clonality: Polyclonal
Clone: I682
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR