Anti-Oct-3 Rabbit Polyclonal Antibody [clone: Q306]

Lieferant: Bioworld Technology
BS3359
BWRLBS3359EA 262 EUR
BWRLBS3359
Anti-Oct-3 Rabbit Polyclonal Antibody [clone: Q306]
Antikörper
Synthetic peptide, corresponding to amino acids 281-330 of Human Oct-3.

OCT3 (Q306) pAb detects endogenous levels of OCT3 protein.

Type: Primary
Antigen: Oct-3
Clonality: Polyclonal
Clone: Q306
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR