Anti-Pmp24 Rabbit Polyclonal Antibody [clone: L27]

Lieferant: Bioworld Technology
BS3366
BWRLBS3366EA 262 EUR
BWRLBS3366
Anti-Pmp24 Rabbit Polyclonal Antibody [clone: L27]
Antikörper
Synthetic peptide, corresponding to the N-terminual of Human Pmp24.

Pmp24 (L27) pAb detects endogenous levels of Pmp24 protein.

Type: Primary
Antigen: Pmp24
Clonality: Polyclonal
Clone: L27
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR