Anti-Kv9.2 Rabbit Polyclonal Antibody [clone: I229]

Lieferant: Bioworld Technology
BS3371
BWRLBS3371EA 262 EUR
BWRLBS3371
Anti-Kv9.2 Rabbit Polyclonal Antibody [clone: I229]
Antikörper
Synthetic peptide, corresponding to amino acids 198-245 of Human KV9.2.

Kv9.2 (I229) pAb detects endogenous levels of Kv9.2 protein.

Type: Primary
Antigen: Kv9.2
Clonality: Polyclonal
Clone: I229
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR