Anti-mGluR-2/3 Rabbit Polyclonal Antibody [clone: N867pAb ]
Lieferant: Bioworld Technology
BS3430
BWRLBS3430EA 262 EUR
BWRLBS3430
Anti-mGluR-2/3 Rabbit Polyclonal Antibody [clone: N867pAb ]
Antikörper
Synthetic peptide, corresponding to amino acids 836-886 of Human mGluR-2.
mGluR-2/3 (N867) pAb detects endogenous levels of mGluR-2/3 protein
Type: Primary
Antigen: mGluR-2/3
Clonality: Polyclonal
Clone: N867pAb
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat