Anti-FucT-IV Rabbit Polyclonal Antibody [clone: E71]
Lieferant: Bioworld Technology
BS3444
BWRLBS3444EA 262 EUR
BWRLBS3444
Anti-FucT-IV Rabbit Polyclonal Antibody [clone: E71]
Antikörper
Synthetic peptide, corresponding to amino acids 45-90 of Human FucT-IV.
FucT-IV (E71) pAb detects endogenous levels of FucT-IV protein.
Type: Primary
Antigen: FucT-IV
Clonality: Polyclonal
Clone: E71
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat