Anti-BMP-5 Rabbit Polyclonal Antibody [clone: L163]

Lieferant: Bioworld Technology
BS3455
BWRLBS3455EA 262 EUR
BWRLBS3455
Anti-BMP-5 Rabbit Polyclonal Antibody [clone: L163]
Antikörper
Synthetic peptide, corresponding to amino acids 130-180 of Human BMP-5.

BMP-5 (L163) pAb detects endogenous levels of BMP-5 protein.

Type: Primary
Antigen: BMP-5
Clonality: Polyclonal
Clone: L163
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR