Anti-IL-20 Rabbit Polyclonal Antibody [clone: D61]

Lieferant: Bioworld Technology
BS3464
BWRLBS3464EA 262 EUR
BWRLBS3464
Anti-IL-20 Rabbit Polyclonal Antibody [clone: D61]
Antikörper
Synthetic peptide, corresponding to amino acids 30-78 of Human IL-20.

IL-20 (D61) pAb detects endogenous levels of IL-20 protein.

Type: Primary
Antigen: IL-20
Clonality: Polyclonal
Clone: D61
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR