Anti-Connexin 45 Rabbit Polyclonal Antibody [clone: N361]

Lieferant: Bioworld Technology
BS3470
BWRLBS3470EA 262 EUR
BWRLBS3470
Anti-Connexin 45 Rabbit Polyclonal Antibody [clone: N361]
Antikörper
Synthetic peptide, corresponding to amino acids 350-400 of Human Connexin 45.

Connexin 45 (N361) pAb detects endogenous levels of Connexin 45 protein.

Type: Primary
Antigen: Connexin 45
Clonality: Polyclonal
Clone: N361
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR