Anti-IL-12B Rabbit Polyclonal Antibody [clone: V277]

Lieferant: Bioworld Technology
BS3480
BWRLBS3480EA 262 EUR
BWRLBS3480
Anti-IL-12B Rabbit Polyclonal Antibody [clone: V277]
Antikörper
Synthetic peptide, corresponding to amino acids 245-291 of Human IL-12B.

IL-12B (V277) pAb detects endogenous levels of IL-12B protein.

Type: Primary
Antigen: IL-12B
Clonality: Polyclonal
Clone: V277
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR