Anti-ABCG2 Rabbit Polyclonal Antibody [clone: N338]

Lieferant: Bioworld Technology
BS3482
BWRLBS3482EA 262 EUR
BWRLBS3482
Anti-ABCG2 Rabbit Polyclonal Antibody [clone: N338]
Antikörper
Synthetic peptide, corresponding to amino acids 302-356 of Human ABCG2.

ABCG2 (N338) pAb detects endogenous levels of ATP-binding cassette sub-family G member 2 protein.

Type: Primary
Antigen: ABCG2
Clonality: Polyclonal
Clone: N338
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR