Anti-HIF-1alpha Rabbit Polyclonal Antibody [clone: K377]
Lieferant: Bioworld Technology
BS3514
BWRLBS3514EA 262 EUR
BWRLBS3514
Anti-HIF-1alpha Rabbit Polyclonal Antibody [clone: K377]
Antikörper
Synthetic peptide, corresponding to amino acids 345-398 of Human HIF-1alpha.
HIF-1? (K377) pAb detects endogenous levels of HIF-1? protein.
Type: Primary
Antigen: HIF-1alpha
Clonality: Polyclonal
Clone: K377
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat