Anti-FoxO3A Rabbit Polyclonal Antibody [clone: P247]
Lieferant: Bioworld Technology
BS3574
BWRLBS3574EA 262 EUR
BWRLBS3574
Anti-FoxO3A Rabbit Polyclonal Antibody [clone: P247]
Antikörper
Synthetic peptide, corresponding to amino acids 221-270 of Human FoxO3A.
FoxO3A (P247) pAb detects endogenous levels of FoxO3A protein.
Type: Primary
Antigen: FoxO3A
Clonality: Polyclonal
Clone: P247
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat