Anti-FAK Rabbit Polyclonal Antibody [clone: I855]

Lieferant: Bioworld Technology
BS3583
BWRLBS3583EA 262 EUR
BWRLBS3583
Anti-FAK Rabbit Polyclonal Antibody [clone: I855]
Antikörper
Synthetic peptide, corresponding to amino acids 830-880 of Human FAK.

FAK (I855) pAb detects endogenous levels of FAK protein.

Type: Primary
Antigen: FAK
Clonality: Polyclonal
Clone: I855
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR