Anti-GluR1 Rabbit Polyclonal Antibody [clone: L844]

Lieferant: Bioworld Technology
BS3656
BWRLBS3656EA 262 EUR
BWRLBS3656
Anti-GluR1 Rabbit Polyclonal Antibody [clone: L844]
Antikörper
Synthetic peptide, corresponding to amino acids 830-880 of Human GluR1.

GluR1 (L844) pAb detects endogenous levels of GluR1 protein.

Type: Primary
Antigen: GluR1
Clonality: Polyclonal
Clone: L844
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR