Anti-GluR1 Rabbit Polyclonal Antibody [clone: S857]

Lieferant: Bioworld Technology
BS3657
BWRLBS3657EA 262 EUR
BWRLBS3657
Anti-GluR1 Rabbit Polyclonal Antibody [clone: S857]
Antikörper
Synthetic peptide, corresponding to amino acids 830-880 of Human GluR1.

GluR1 (S857) pAb detects endogenous levels of GluR1 protein.

Type: Primary
Antigen: GluR1
Clonality: Polyclonal
Clone: S857
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR