Anti-Integrin beta3 Rabbit Polyclonal Antibody [clone: S778]

Lieferant: Bioworld Technology
BS3660
BWRLBS3660EA 262 EUR
BWRLBS3660
Anti-Integrin beta3 Rabbit Polyclonal Antibody [clone: S778]
Antikörper
Synthetic peptide, corresponding to amino acids 751-800 of Human Integrin beta3.

Integrin ?3 (S778) pAb detects endogenous levels of Integrin ?3 protein.

Type: Primary
Antigen: Integrin beta3
Clonality: Polyclonal
Clone: S778
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR