Anti-BMP-7 Rabbit Polyclonal Antibody [clone: E173]

Lieferant: Bioworld Technology
BS3674
BWRLBS3674EA 262 EUR
BWRLBS3674
Anti-BMP-7 Rabbit Polyclonal Antibody [clone: E173]
Antikörper
Synthetic peptide, corresponding to amino acids 140-190 of Human BMP-7.

BMP-7 (E173) pAb detects endogenous levels of BMP-7 protein.

Type: Primary
Antigen: BMP-7
Clonality: Polyclonal
Clone: E173
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR