Anti-BMP-8 Rabbit Polyclonal Antibody [clone: E90]

Lieferant: Bioworld Technology
BS3677
BWRLBS3677EA 262 EUR
BWRLBS3677
Anti-BMP-8 Rabbit Polyclonal Antibody [clone: E90]
Antikörper
Synthetic peptide, corresponding to amino acids 70-120 of Human BMP-8.

BMP-8 (E90) pAb detects endogenous levels of BMP-8A and BMP-8B protein.

Type: Primary
Antigen: BMP-8
Clonality: Polyclonal
Clone: E90
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR