Anti-BMP-3 Rabbit Polyclonal Antibody [clone: E306]

Lieferant: Bioworld Technology
BS3683
BWRLBS3683EA 262 EUR
BWRLBS3683
Anti-BMP-3 Rabbit Polyclonal Antibody [clone: E306]
Antikörper
Synthetic peptide, corresponding to amino acids 261-310 of Human BMP-3.

BMP-3 (E306) pAb detects endogenous levels of BMP-3 protein.

Type: Primary
Antigen: BMP-3
Clonality: Polyclonal
Clone: E306
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR