Anti-TYK2 Rabbit Polyclonal Antibody [clone: V1048]

Lieferant: Bioworld Technology
BS3713
BWRLBS3713EA 262 EUR
BWRLBS3713
Anti-TYK2 Rabbit Polyclonal Antibody [clone: V1048]
Antikörper
Synthetic peptide, corresponding to amino acids 1010-1065 of Human TYK2.

Tyk 2 (V1048) pAb detects endogenous levels of Tyk 2 protein.

Type: Primary
Antigen: TYK2
Clonality: Polyclonal
Clone: V1048
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR