Anti-Rad51 Rabbit Polyclonal Antibody [clone: T309]

Lieferant: Bioworld Technology
BS3729
BWRLBS3729EA 262 EUR
BWRLBS3729
Anti-Rad51 Rabbit Polyclonal Antibody [clone: T309]
Antikörper
Synthetic peptide, corresponding to amino acids 280-330 of Human Rad51.

Rad51 (T309) pAb detects endogenous levels of Rad51 protein.

Type: Primary
Antigen: Rad51
Clonality: Polyclonal
Clone: T309
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR