Anti-ILK Rabbit Polyclonal Antibody [clone: P240]
Lieferant: Bioworld Technology
BS3743
BWRLBS3743EA 262 EUR
BWRLBS3743
Anti-ILK Rabbit Polyclonal Antibody [clone: P240]
Antikörper
Synthetic peptide, corresponding to amino acids 212-258 of Human ILK.
ILK (P240) pAb detects endogenous levels of ILK protein.
Type: Primary
Antigen: ILK
Clonality: Polyclonal
Clone: P240
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat