Anti-c-Myb Rabbit Polyclonal Antibody [clone: I526]

Lieferant: Bioworld Technology
BS3769
BWRLBS3769EA 262 EUR
BWRLBS3769
Anti-c-Myb Rabbit Polyclonal Antibody [clone: I526]
Antikörper
Synthetic peptide, corresponding to amino acids 491-540 of Human c-Myb.

c-Myb (I526) pAb detects endogenous levels of c-Myb protein.

Type: Primary
Antigen: c-Myb
Clonality: Polyclonal
Clone: I526
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR