Anti-PSMD11 Rabbit Polyclonal Antibody [clone: R310]

Lieferant: Bioworld Technology
BS3780
BWRLBS3780EA 262 EUR
BWRLBS3780
Anti-PSMD11 Rabbit Polyclonal Antibody [clone: R310]
Antikörper
Synthetic peptide, corresponding to amino acids 281-330 of Human PSMD11.

PSMD11 (R310) pAb detects endogenous levels of PSMD11 protein.

Type: Primary
Antigen: PSMD11
Clonality: Polyclonal
Clone: R310
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR