Anti-PSMD12 Rabbit Polyclonal Antibody [clone: I186]

Lieferant: Bioworld Technology
BS3781
BWRLBS3781EA 262 EUR
BWRLBS3781
Anti-PSMD12 Rabbit Polyclonal Antibody [clone: I186]
Antikörper
Synthetic peptide, corresponding to amino acids 161-210 of Human PSMD12.

PSMD12 (I186) pAb detects endogenous levels of PSMD12 protein.

Type: Primary
Antigen: PSMD12
Clonality: Polyclonal
Clone: I186
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR