Anti-MRP-S9 Rabbit Polyclonal Antibody [clone: F132]

Lieferant: Bioworld Technology
BS3783
BWRLBS3783EA 262 EUR
BWRLBS3783
Anti-MRP-S9 Rabbit Polyclonal Antibody [clone: F132]
Antikörper
Synthetic peptide, corresponding to amino acids 100-150 of Human MRP-S9.

MRP-S9 (F132) pAb detects endogenous levels of MRP-S9 protein.

Type: Primary
Antigen: MRP-S9
Clonality: Polyclonal
Clone: F132
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR