Anti-ACAT1 Rabbit Polyclonal Antibody [clone: K266]
Lieferant: Bioworld Technology
BS3796
BWRLBS3796EA 262 EUR
BWRLBS3796
Anti-ACAT1 Rabbit Polyclonal Antibody [clone: K266]
Antikörper
Synthetic peptide, corresponding to amino acids 235-286 of Human ACAT1.
ACAT1 (K266) pAb detects endogenous levels of ACAT1 protein.
Type: Primary
Antigen: ACAT1
Clonality: Polyclonal
Clone: K266
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat