Anti-CPM Rabbit Polyclonal Antibody [clone: L113]
Lieferant: Bioworld Technology
BS3827
BWRLBS3827EA 262 EUR
BWRLBS3827
Anti-CPM Rabbit Polyclonal Antibody [clone: L113]
Antikörper
Synthetic peptide, corresponding to amino acids 80-130 of Human CPM.
CPM (L113) pAb detects endogenous levels of CPM protein.
Type: Primary
Antigen: CPM
Clonality: Polyclonal
Clone: L113
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat