Anti-CEP290 Rabbit Polyclonal Antibody [clone: F812]

Lieferant: Bioworld Technology
BS3831
BWRLBS3831EA 262 EUR
BWRLBS3831
Anti-CEP290 Rabbit Polyclonal Antibody [clone: F812]
Antikörper
Synthetic peptide, corresponding to amino acids 781-830 of Human CEP290.

CEP290 (F812) pAb detects endogenous levels of CEP290 protein.

Type: Primary
Antigen: CEP290
Clonality: Polyclonal
Clone: F812
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR