Anti-C9 Rabbit Polyclonal Antibody [clone: H216]

Lieferant: Bioworld Technology
BS3841
BWRLBS3841EA 262 EUR
BWRLBS3841
Anti-C9 Rabbit Polyclonal Antibody [clone: H216]
Antikörper
Synthetic peptide, corresponding to amino acids 171-220 of Human C9.

C9 (H216) pAb detects endogenous levels of Complement component C9 protein.

Type: Primary
Antigen: C9
Clonality: Polyclonal
Clone: H216
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR