Anti-IFI-16 Rabbit Polyclonal Antibody [clone: N772]

Lieferant: Bioworld Technology
BS3865
BWRLBS3865EA 262 EUR
BWRLBS3865
Anti-IFI-16 Rabbit Polyclonal Antibody [clone: N772]
Antikörper
Synthetic peptide, corresponding to amino acids 740-795 of Human IFI-16.

IFI-16 (N772) pAb detects endogenous levels of IFI-16 protein.

Type: Primary
Antigen: IFI-16
Clonality: Polyclonal
Clone: N772
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR