Anti-HLA-H Rabbit Polyclonal Antibody [clone: I104]

Lieferant: Bioworld Technology
BS3876
BWRLBS3876EA 262 EUR
BWRLBS3876
Anti-HLA-H Rabbit Polyclonal Antibody [clone: I104]
Antikörper
Synthetic peptide, corresponding to amino acids 72-126 of Human HLA-H.

HLA-H (I104) pAb detects endogenous levels of HLA-H protein.

Type: Primary
Antigen: HLA-H
Clonality: Polyclonal
Clone: I104
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR