Anti-Renin Rabbit Polyclonal Antibody [clone: L238]

Lieferant: Bioworld Technology
BS3925
BWRLBS3925EA 262 EUR
BWRLBS3925
Anti-Renin Rabbit Polyclonal Antibody [clone: L238]
Antikörper
Synthetic peptide, corresponding to amino acids 211-260 of Human Renin.

Renin (L238) pAb detects endogenous levels of Renin protein.

Type: Primary
Antigen: Renin
Clonality: Polyclonal
Clone: L238
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR