Anti-p-AKT Rabbit Polyclonal Antibody [clone: T308]

Lieferant: Bioworld Technology
BS4009
BWRLBS4009EA 284 EUR
BWRLBS4009
Anti-p-AKT Rabbit Polyclonal Antibody [clone: T308]
Antikörper
Synthetic phosphopeptide derived from human AKT1 around the phosphorylation site of Threonine 308.

p-AKT (T308) pAb detects endogenous levels of AKT1 only when phosphorylated at Thr308. This antibody also recognizes AKT2 and AKT3 when phosphorylated at the corresponding residues.

Type: Primary
Antigen: p-AKT
Clonality: Polyclonal
Clone: T308
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR