Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: T161]

Lieferant: Bioworld Technology
BS4033
BWRLBS4033EA 284 EUR
BWRLBS4033
Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: T161]
Antikörper
Synthetic phosphopeptide derived from human Cdk1/Cdc2 around the phosphorylation site of Serine 795.

p-Cdk1/Cdc2 (T161) pAb detects endogenous levels of p-Cdk1/Cdc2 protein only when phosphorylated at Thr161.

Type: Primary
Antigen: p-Cdk1/Cdc2
Clonality: Polyclonal
Clone: T161
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR