Anti-p-JunB Rabbit Polyclonal Antibody [clone: S259]

Lieferant: Bioworld Technology
BS4110
BWRLBS4110EA 284 EUR
BWRLBS4110
Anti-p-JunB Rabbit Polyclonal Antibody [clone: S259]
Antikörper
Synthetic phosphopeptide derived from human JunB around the phosphorylation site of Serine 259.

p-JunB (S259) pAb detects endogenous levels of JunB protein only when phosphorylated at Ser259.

Type: Primary
Antigen: p-JunB
Clonality: Polyclonal
Clone: S259
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR